Tributylgermanium 97
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Tributylgermanium hydride, Tri-n-butylgermanium hydride
Purity: 97+%
CAS No.: 998-39-0
Molecular Formula: [CH3(CH2)3]3GeH
Formula Weight: 244.95
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 0.92
Refractive Index: 1.4508
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless to pale yellow
Boiling Point: 123°/20mm
MDL No.: MFCD00077987
Reduction of benzylic chlorides
Formel:
C₁₂H₂₈Ge Molekylvægt: 244.96 g/mol Kogepunkt: 123 °C (20 mmHg) Massefylde: 0.92 Flammepunkt: 113 °C (235 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00077987 CAS-nummer: 998-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...