Diethylsilan ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98+%
CAS No.: 542-91-6
Molecular Formula: C4H12Si
Formula Weight: 88.23
Flash Point: -20°(-4°F)
Refractive Index: 1.3920
Odour: Mild
Melting Point: -132°
Boiling Point: 56°
Beilstein Reference: 1730902
Merck Reference: 14,3128
UN No.: 1993
MDL No.: MFCD00039877
Formel:
C₄H₁₂Si Kogepunkt: 56 °C Smeltepunkt: –132 °C Massefylde: 0,684 Flammepunkt: –20 °C (–4 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00039877 CAS-nummer: 542-91-6 EINECS: 208-834-9 UN nummer: 1993 ADR: 3,II Merck Index: 14,3128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...