Dibutyltinoxid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

179361000 179365000
ACRO179361000EA 310 DKK
ACRO179361000 ACRO179365000
Dibutyltinoxid 98%
Dibutyltinoxid
Formel: C₈H₁₈OSn
Smeltepunkt: >300 °C
Massefylde: 1,6 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001992
CAS-nummer: 818-08-6
EINECS: 212-449-1
UN nummer: 3146
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White Powder or granular powder
Infrared spectrum Conforms
Chloride (Cl) ≤0.2 %
Tin (Sn) ≥47 %
Volatile matter ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR