Octyltrimethoxysilan ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Trimethoxy(octyl)silane
42698.06
42698.14
42698.22
42698.06EA
465
DKK
42698.06
42698.14
42698.22
Octyltrimethoxysilan ≥97%
Octyltrimethoxysilane
Purity: 97+%
CAS No.: 3069-40-7
Molecular Formula: C11H26O3Si
Formula Weight: 234.41
Flash Point: 102°(216°F)
Physical Form: Liquid
Density: 0.907
Refractive Index: 1.417
Sensitivity: Moisture Sensitive
Appearance: Colourless to yellow
Boiling Point: 75°/0.1mm
MDL No.: MFCD00043053
Formel:
C₁₁H₂₆O₃Si Kogepunkt: 75 °C (0,1 mmHg) Massefylde: 0,907 Flammepunkt: 102 °C (216 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00043053 CAS-nummer: 3069-40-7 EINECS: 221-338-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...