Octadecyltriethoxysilan 98% (n-isomer ≥ 85%)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Triethoxy(octadecyl)silane
L06366.09
L06366.18
L06366.09EA
536
DKK
L06366.09
L06366.18
Octadecyltriethoxysilan 98% (n-isomer ≥ 85%)
Octadecyltriethoxysilane
Purity: 98%
CAS No.: 7399-00-0
Molecular Formula: CH3(CH2)17Si(OCH2CH3)3
Formula Weight: 416.77
Flash Point: 195°(383°F)
Refractive Index: 1.4380
Boiling Point: 165-169°/0.2mm
Beilstein Reference: 7025163
MDL No.: MFCD00039875
Formel:
C₂₄H₅₂O₃Si Kogepunkt: 165…169 °C (0,2 mmHg) Massefylde: 0,870 Flammepunkt: 195 °C (383 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00039875 CAS-nummer: 7399-00-0 EINECS: 230-995-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...