Methyltrichlorsilan for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Trichloro(methyl)silane
M-Clarity™ quality level = MQ200
8.08255.0250 8.08255.1000
8.08255.0250EA 308 DKK
8.08255.0250 8.08255.1000
Methyltrichlorsilan for synthesis, Sigma-Aldrich®
Methyltrichlorsilan
Formel: CH₃Cl₃Si
Kogepunkt: 66 °C (1013 hPa)
Smeltepunkt: –90 °C
Massefylde: 1,27 g/cm³ (20 °C)
Flammepunkt: 1 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000481
CAS-nummer: 75-79-6
EINECS: 200-902-6
UN nummer: 1250
ADR: 3 (8),II
REACH: 01-2119475226-36
Bestil nu
Specifikationer Testresultater
Solubility (20 °C) (decomposition)
Melting Point -90 °C
Molar Mass 149.47 g/mol
Boiling Point 66 °C
Vapor Pressure 189 hPa (20 °C)
Flash Point -1 °C
Refractive Index 1.411 (20 °C, 589 nm)
Explosion Limit 5.5 - 10.4 %(V)
Density 1.27 g/cm3 (20 °C)
pH (H2O) acidic,Hydrolysis
Ignition Point 500 °C