Tin(II)sulfat 95%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Tin(II) sulphate, Tin sulphate, Stannous sulphate
223661000 223665000 223660025
ACRO223661000EA 345 DKK
ACRO223661000 ACRO223665000 ACRO223660025
Tin(II)sulfat 95%
Tin(II)sulfat
Formel: O₄SSn
Molekylvægt: 214.77 g/mol
Smeltepunkt: 360 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011246
CAS-nummer: 7488-55-3
EINECS: 231-302-2
Merck Index: 12,08947
Bestil nu
Specifikationer Testresultater
Appearance White to off-white Powder or crystals and/or lumps
Titration Iodimetric ≥94.0 %