Tetramethoxysilan 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methylsilikat

203820250 203821000 203825000 203820025
ACRO203820250EA 253 DKK
ACRO203820250 ACRO203821000 ACRO203825000 ACRO203820025
Tetramethoxysilan 99%
Tetramethoxysilane
Formel: C₄H₁₂O₄Si
Molekylvægt: 152.22 g/mol
Kogepunkt: 121
Smeltepunkt: –2 °C
Massefylde: 1.03 g/cm³
Flammepunkt: 26
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008341
CAS-nummer: 681-84-5
EINECS: 211-656-4
UN nummer: 2606
ADR: 6.1 I

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3670 to 1.3700 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR