Tetrabutoxysilan 97
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Tetrabutyl orthosilicate
L03141.14
L03141.22
L03141.14EA
651
DKK
L03141.14
L03141.22
Tetrabutoxysilan 97
Tetrabutoxysilane
Purity: 97%
CAS No.: 4766-57-8
Molecular Formula: C16H36O4Si
Formula Weight: 320.55
Flash Point: 135°(275°F)
Refractive Index: 1.4130
Odour: Characteristic
Melting Point:
Formel:
C₁₆H₃₆O₄Si Molekylvægt: 320.54 g/mol Kogepunkt: 277 °C Smeltepunkt: < –80 °C Massefylde: 0.899 Flammepunkt: 135 °C (275 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009432 CAS-nummer: 4766-57-8 EINECS: 225-305-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...