Triethoxysilan 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B22063.09
B22063.18
B22063.30
B22063.09EA
765
DKK
B22063.09
B22063.18
B22063.30
Triethoxysilan 96%
Triethoxysilane
Purity: 96%
CAS No.: 998-30-1
Molecular Formula: C6H16O3Si
Formula Weight: 164.28
Flash Point: 26°(78°F)
Refractive Index: 1.3770
Melting Point: -170°
Boiling Point: 134-135°
Beilstein Reference: 1738989
UN No.: 3384
MDL No.: MFCD00009061
Formel:
C₆H₁₆O₃Si Kogepunkt: 134…135 °C Smeltepunkt: –170 °C Massefylde: 0,885 Flammepunkt: 26 °C (78 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009061 CAS-nummer: 998-30-1 EINECS: 213-650-7 UN nummer: 3489 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...