Tin(II)fluorid 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tin difluoride

308501000 308500050 308505000
ACRO308501000EA 682 DKK
ACRO308501000 ACRO308500050 ACRO308505000
Tin(II)fluorid 99%
Tin(II)fluorid
Formel: SnF₂
Molekylvægt: 156.71 g/mol
Kogepunkt: 850 °C (1013 hPa)
Smeltepunkt: 215 °C
Massefylde: 4.57
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042540
CAS-nummer: 7783-47-3
EINECS: 231-999-3
UN nummer: 2923
ADR: 8 III
Merck Index: 12,08940

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder or crystals
Gravimetry ≥98.5 % (ex Sn)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR