Indium(III)sulfid ≥99.995% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diindium trisulphide, Indium trisulphide

44836.09 44836.18
44836.09EA 3310 DKK
44836.09 44836.18
Indium(III)sulfid ≥99.995% (metals basis)
Indium(III) sulphide

Purity: 99.995% CAS No.: 12030-24-9 Molecular Formula: In2S3 Formula Weight: 325.83 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Red-orangw Odour: Like rotten eggs Melting Point: 1050° MDL No.: MFCD00011061

Formel: In₂S₃
Molekylvægt: 325.84 g/mol
Smeltepunkt: 1050 °C
Massefylde: 4.89 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011061
CAS-nummer: 12030-24-9
EINECS: 234-742-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR