Aluminumoxid, γ-α-fase ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

14558.14 14558.22 14558.36
14558.14EA 545 DKK
14558.14 14558.22 14558.36
Aluminumoxid, γ-α-fase ≥99.99% (metals basis)
Aluminumoxid, γ-α-fase

Purity: 99.99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.3 Micron Powder, S.A. 15m2/g Odour: Odourless Merck Reference: 14,356 MDL No.: MFCD00003424

Formel: Al₂O₃
Molekylvægt: 101.96 g/mol
Kogepunkt: 2980
Smeltepunkt: 2045 °C
Massefylde: 3.97 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003424
CAS-nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR