Indium(III)iodid, anhydrous ≥99.999% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

11854.04 11854.09 11854.18
11854.04EA 1320 DKK
11854.04 11854.09 11854.18
Indium(III)iodid, anhydrous ≥99.999% (metals basis)
Indium(III) iodide

Purity: 99.999% CAS No.: 13510-35-5 Molecular Formula: I3In Formula Weight: 495.53 Physical Form: Crystalline Density: 4.69 Appearance: Yellow to brown Melting Point: 210° UN No.: 3260 MDL No.: MFCD00016151

Formel: I₃In
Molekylvægt: 495.53 g/mol
Kogepunkt: 500
Smeltepunkt: 210 °C
Massefylde: 4.69
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016151
CAS-nummer: 13510-35-5
EINECS: 236-839-6
UN nummer: 3260
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR