Indium(III)bromid 99.99% (trace metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Indium tribromide

389030100 389030500
ACRO389030100EA 1560 DKK
ACRO389030100 ACRO389030500
Indium(III)bromid 99.99% (trace metals basis)
Indium(III) bromide
Formel: Br₃In
Molekylvægt: 354.53 g/mol
Kogepunkt: 656
Smeltepunkt: 220 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011057
CAS-nummer: 13465-09-3
EINECS: 236-692-8

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder or crystals or granules and/or chunks
Total trace metal impurities ≤100 ppm
Indium (In) 31.7 to 33.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR