Bariumsulfat, EMPROVE® ESSENTIAL Ph. Eur., USP, BP, extra pure for X-ray diagnosis, suitable for use as excipient, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.01750.5000 1.01750.9050 1.01750.9025
1.01750.5000EA 5610 DKK
1.01750.5000 1.01750.9050 1.01750.9025
Bariumsulfat, EMPROVE® ESSENTIAL Ph. Eur., USP, BP, extra pure for X-ray diagnosis, suitable for use as excipient, SAFC®
Barium sulphate
Formel: BaSO₄
Molekylvægt: 233.39 g/mol
Kogepunkt: 1600
Smeltepunkt: 1345
Massefylde: 4.5 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003455
CAS-nummer: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997