Strontiumsulfid ≥99.9% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
89895.09
89895.18
89895.09EA
1880
DKK
89895.09
89895.18
Strontiumsulfid ≥99.9% (metals basis)
Strontium sulphide
Purity: 99.9%
CAS No.: 1314-96-1
Molecular Formula: SrS
Formula Weight: 119.68
Physical Form: Powder
Density: 3.70
Appearance: Grey
Odour: Like rotten eggs
Melting Point:
Formel:
SSr Smeltepunkt: >2000 °C Massefylde: 3,70 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00049560 CAS-nummer: 1314-96-1 EINECS: 215-249-2 UN nummer: 3134 ADR: 4.3,III Merck Index: 14,8849 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...