Barium manganat tech. 90%, Technical Grade
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Barium manganate(VI)
L13172.06
L13172.14
L13172.06EA
317
DKK
L13172.06
L13172.14
Barium manganat tech. 90%, Technical Grade
Barium manganat
Purity: 90%
CAS No.: 7787-35-1
Molecular Formula: BaMnO4
Formula Weight: 256.28
Merck Reference: 14,981
UN No.: 3087
MDL No.: MFCD00011514
Formel:
BaMnO₄ Massefylde: 4,850 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011514 CAS-nummer: 7787-35-1 EINECS: 232-109-6 UN nummer: 3087 ADR: 5.1,III Merck Index: 14,981 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...