Calciumcarbonat 99,0 ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Marble
33295.22
33295.36
33295.22EA
394
DKK
33295.22
33295.36
Calciumcarbonat 99,0 ACS
Calcium carbonate
Purity: 99.0% min
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odour: Odourless
Melting Point: 800° dec.
Merck Reference: 14,1657
MDL No.: MFCD00010906
Formel:
CaCO₃ Molekylvægt: 100.09 g/mol Smeltepunkt: 800 °C (dec.) Massefylde: 2.930 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010906 CAS-nummer: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...