Dikaliumperoxodisulfat ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate

A15310.0E A15310.36
A15310.0EEA 2420 DKK
A15310.0E A15310.36
Dikaliumperoxodisulfat ≥97%
Dikaliumperoxodisulfat

Purity: 97% CAS No.: 7727-21-1 Molecular Formula: K2O8S2 Formula Weight: 270.33 Odour: Odourless Melting Point: 100° dec. UN No.: 1492 MDL No.: MFCD00011386

Formel: K₂S₂O₈
Molekylvægt: 270.32 g/mol
Smeltepunkt: 100 °C (dec.)
Massefylde: 2.477
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011386
CAS-nummer: 7727-21-1
EINECS: 231-781-8
UN nummer: 1492
ADR: 5.1 III
Merck Index: 13,07741

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR