Kaliumdisulfit
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
7995.1
7995.2
ROTH7995.1EA
0
DKK
ROTH7995.1
ROTH7995.2
Kaliumdisulfit
Potassium disulphite
Formel:
K₂O₅S₂ Molekylvægt: 222.33 g/mol Smeltepunkt: 190 Massefylde: 1.2 g/cm³ (20 °C) |
MDL Number:
MFCD00167605 CAS-nummer: 16731-55-8 EINECS: 240-795-3 Merck Index: 13,07729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...