Natriumdihydrogenphosphat
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
T878.1 T878.2
ROTHT878.1EA 257.32 DKK
ROTHT878.1 ROTHT878.2
Natriumdihydrogenphosphat
Natriumdihydrogenphosphat
Formel: NaH₂PO₄
Molekylvægt: 119.98 g/mol
Smeltepunkt: ∼650 °C
Massefylde: 0.9996 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003527
CAS-nummer: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806