Kaliumdihydrogenorthophosphat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

P018.1 P018.2 P018.5
ROTHP018.1EA 190.19 DKK
ROTHP018.1 ROTHP018.2 ROTHP018.5
Kaliumdihydrogenorthophosphat
Potassium dihydrogen phosphate
Formel: KH₂PO₄
Molekylvægt: 136.09 g/mol
Kogepunkt: ∼100 °C (1013 hPa)
Smeltepunkt: 253
Massefylde: 2.3 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011401
CAS-nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR