Natriumperchlorat ≥98.0% ACS, Fluka™
Leverandør: Honeywell Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
410241-2.5KG
410241-500G
410241-25KG
410241-2.5KGEA
8690
DKK
410241-2.5KG
410241-500G
HONC410241-25KG
Natriumperchlorat ≥98.0% ACS, Fluka™
Sodium perchlorate
Formel:
ClNaO₄ Smeltepunkt: 469 °C (dec.) Massefylde: 2,52 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003480 CAS-nummer: 7601-89-0 EINECS: 231-511-9 UN nummer: 1502 ADR: 5.1,II Merck Index: 13,08726 |
Specifikationer Testresultater
Assay | 98.0 - 102.0 % |
pH (5 %, 25 °C) | 6.0 - 8.0 |
Water insoluble matter | Max. 0.005 % |
Calcium (Ca) | Max. 0.02 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.05 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.003 % |
Sulfate (SO₄) | Max. 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...