1,1,3,3-Tetramethylguanidin for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N,N',N'-Tetramethylguanidine

M-Clarity™ quality level = MQ200

8.21897.0100 8.21897.0250 8.21897.9029 8.21897.9010
8.21897.0100EA 499 DKK
8.21897.0100 8.21897.0250 8.21897.9029 8.21897.9010
1,1,3,3-Tetramethylguanidin for synthesis, Sigma-Aldrich®
1,1,3,3-Tetramethylguanidin
Formel: C₅H₁₃N₃
Molekylvægt: 115.18 g/mol
Kogepunkt: 160
Smeltepunkt: –30 °C
Massefylde: 0.92 g/cm³ (20 °C)
Flammepunkt: 54
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008323
CAS-nummer: 80-70-6
EINECS: 201-302-7
UN nummer: 2920
ADR: 8 (3) II

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Molar Mass 115.17 g/mol
Boiling Point 162 °C (1013 hPa)
Vapor Pressure 0.26 hPa (25 °C)
Flash Point 50 °C
Refractive Index 1.4659 (20 °C, 589 nm)
Explosion Limit 1.0 - 7.5 %(V)
Density 0.92 g/cm3 (20 °C)
pH 12.7 (10 g/l, H2O, 25 °C)
Ignition Point 350 °C
Hill Formula C5H13N3
Chemical Formula [(CH3)2N]2C=NH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR