5-Methyl-2-furaldehyd 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

159920250 159921000
ACRO159920250EA 276 DKK
ACRO159920250 ACRO159921000
5-Methyl-2-furaldehyd 98+%
5-Methyl-2-furaldehyd
Formel: C₆H₆O₂
Kogepunkt: 89 °C (1013 hPa)
Massefylde: 1,1 g/cm³
Flammepunkt: 72 °C
MDL Number: MFCD00003232
CAS-nummer: 620-02-0
EINECS: 210-622-6

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5320 (20°C, 589 nm)
GC ≥98.0 %
Acid value ≤10 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR