2-Methyl-9,10-anthraquinon 97

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Methylanthraquinone, 2-Methylanthracene-9,10-Dione

A14502.0B A14502.18 A14502.30
A14502.0BEA 4500 DKK
A14502.0B A14502.18 A14502.30
2-Methyl-9,10-anthraquinon 97
2-Methyl-9,10-anthraquinon

Purity: 97% CAS No.: 84-54-8 Molecular Formula: C15H10O2 Formula Weight: 222.24 Flash Point: 209°(408°F) Melting Point: 170-174° Beilstein Reference: 2050523 Merck Reference: 14,6021 MDL No.: MFCD00001235

Formel: C₁₅H₁₀O₂
Molekylvægt: 222.24 g/mol
Kogepunkt: 236…238 °C (10 mmHg)
Smeltepunkt: 170…174 °C
Massefylde: 1.365 g/cm³ (20 °C)
Flammepunkt: 209 °C (408 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001235
CAS-nummer: 84-54-8
EINECS: 201-539-6
UN nummer: 3077
ADR: 9 III
Merck Index: 12,06100

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR