myo-Inositol, white powder cell culture reagent

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

0219468890 0219468850 0219468880 0219468891
ICNA0219468890EA 2630 DKK
ICNA0219468890 ICNA0219468850 ICNA0219468880 ICNA0219468891
myo-Inositol, white powder cell culture reagent
myo-Inositol

Storage: Store at Room Temperature (15-30 °C).
D-myo-Inositol is a component of membrane phospholipids, glycosylphophatidylinositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers. It is a growth factor for animals as well as microorganisms. Myo-Inositol is the most abundant form of polyols that serves as a structural element of secondary messengers in eukaryotic cells. Inositol or cyclohexane-1,2,3,4,5,6-hexol is a chemical compound, a sixfold alcohol (polyol) of cyclohexane. It exists in nine possible stereoisomers, of which the most prominent form, widely occurring in nature, is cis-1,2,3,5-trans-4,6-cyclohexanehexol, or myo-inositol (former name meso-inositol). Inositol is a carbohydrate, though not a classical sugar. It has a taste which has been assayed at half the sweetness of table sugar. Used as a lipotropic agent.
A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers. In addition, inositol serves as an important component of the structural lipids phosphatidylinositol (PI) and its various phosphates, the phosphatidylinositol phosphate (PIP) lipids.


  • Cell Culture Reagent Grade
  • Vapor Density: 6.2 (vs air) (Lit.)
  • Presentation: White Powder

Formel: C₆H₁₂O₆
Kogepunkt: 291 °C (1013 hPa)
Smeltepunkt: 220…230 °C
Massefylde: 1,752 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077932
CAS-nummer: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Bestil nu

Specifikationer Testresultater

Identity Test Passes
FTIR Conforms to Standard
Solubility (10% aq.) Clear Solution
pH (10% aq.) Neutral to Litmus


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR