Dicyclohexylamin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: DCHA

A15671.0F A15671.AE A15671.AP
A15671.0FEA 1190 DKK
A15671.0F A15671.AE A15671.AP
Dicyclohexylamin 98%
Dicyclohexylamin

Purity: 98% CAS No.: 101-83-7 Molecular Formula: C12H23N Formula Weight: 181.32 Flash Point: 99°(210°F) Refractive Index: 1.4842 Melting Point: -2° Boiling Point: 256° Beilstein Reference: 605923 Merck Reference: 14,3095 UN No.: 2565 MDL No.: MFCD00011658

Formel: C₁₂H₂₃N
Molekylvægt: 181.32 g/mol
Kogepunkt: 256 °C
Smeltepunkt: –2 °C
Massefylde: 0.913
Flammepunkt: 99 °C (210 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011658
CAS-nummer: 101-83-7
EINECS: 202-980-7
UN nummer: 2565
ADR: 8 III
Merck Index: 13,03122

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR