7-Methoxycoumarin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
|
Synonymer:
7-Methoxy-2-benzopyrone, 7-Methoxy-2H-chromen-2-one
A15441.06
A15441.14
A15441.22
A15441.06EA
508
DKK
A15441.06
A15441.14
A15441.22
7-Methoxycoumarin ≥98%
7-Methoxycoumarin
Purity: 98+%
CAS No.: 531-59-9
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 117-120°
Beilstein Reference: 141728
Formel:
C₁₀H₈O₃ Molekylvægt: 176.17 g/mol Kogepunkt: 320 Smeltepunkt: 117…120 °C Massefylde: 1.36 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006876 CAS-nummer: 531-59-9 EINECS: 208-513-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...