4,4'-Diiodbiphenyl ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4,4'-Diiodo-1,1'-biphenyl

B21970.14 B21970.22
B21970.14EA 606 DKK
B21970.14 B21970.22
4,4'-Diiodbiphenyl ≥99%
4,4'-Diiodbiphenyl

Purity: 99% CAS No.: 3001-15-8 Molecular Formula: C12H8I2 Formula Weight: 406.00 Melting Point: 205-211° UN No.: 3152 MDL No.: MFCD00001057

Formel: C₁₂H₈I₂
Smeltepunkt: 205…211 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001057
CAS-nummer: 3001-15-8
EINECS: 221-080-5
ADR: 9,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR