Benzhydrylalkohol ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

A12884.0B A12884.18 A12884.30
A12884.0BEA 1700 DKK
A12884.0B A12884.18 A12884.30
Benzhydrylalkohol ≥99%
Benzhydrylalkohol

Purity: 99% CAS No.: 91-01-0 Molecular Formula: C13H12O Formula Weight: 184.24 Flash Point: 160°(320°F) Odour: Odourless Boiling Point: 297-298° Beilstein Reference: 1424379 Merck Reference: 14,1090 MDL No.: MFCD00004488

Formel: C₁₃H₁₂O
Molekylvægt: 184.24 g/mol
Kogepunkt: 297…298 °C
Smeltepunkt: 64…68 °C
Massefylde: 1.054 g/cm³ (80 °C)
Flammepunkt: 160 °C (320 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004488
CAS-nummer: 91-01-0
EINECS: 202-033-8
Merck Index: 12,01121

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR