Phenolphthalein ACS
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
417181000 417185000 417180026
ACRO417181000EA 343 DKK
ACRO417181000 ACRO417185000 ACRO417180026
Phenolphthalein ACS
Phenolphthalein
Formel: C₂₀H₁₄O₄
Molekylvægt: 318.33 g/mol
Smeltepunkt: 258…263 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005913
CAS-nummer: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (colorless) to pH 10 (red)
Clarity of alcohol solution Passes test