Benzophenon for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Diphenylmethanone, Diphenyl ketone

M-Clarity™ quality level = MQ200

8.01801.0005 8.01801.0250 8.01801.1000 8.01801.9050
8.01801.0005EA 171 DKK
8.01801.0005 8.01801.0250 8.01801.1000 8.01801.9050
Benzophenon for synthesis, Sigma-Aldrich®
Benzophenon
Formel: C₁₃H₁₀O
Molekylvægt: 182.22 g/mol
Kogepunkt: 305.4
Smeltepunkt: 47…49 °C
Massefylde: 1.085 g/cm³ (20 °C)
Flammepunkt: 150
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003076
CAS-nummer: 119-61-9
EINECS: 204-337-6
UN nummer: 3077
ADR: 9 III
Merck Index: 13,01099

Bestil nu

Specifikationer Testresultater

Solubility 0.14 g/l (25 °C)
Melting Point 47 - 49 °C
Molar Mass 182.22 g/mol
Bulk Density 700 kg/m3
Boiling Point 304 - 306 °C (1013 hPa)
Vapor Pressure 0.004 hPa (20 °C)
Flash Point 150 °C
Density 1.1 g/cm3 (25 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR