α,4-Dichlor-α-phenyltoluen 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Chlorobenzhydryl chloride, 1-Chloro-4-(chloro(phenyl)methyl)benzene
B22778.14
B22778.22
B22778.36
B22778.14EA
443
DKK
B22778.14
B22778.22
B22778.36
α,4-Dichlor-α-phenyltoluen 97%
α,4-Dichlor-α-phenyltoluen
Purity: 98%
CAS No.: 134-83-8
Molecular Formula: C13H10Cl2
Formula Weight: 237.13
Flash Point: 194°(381°F)
Refractive Index: 1.6030
Boiling Point: 159-160°/2mm
Beilstein Reference: 2050236
UN No.: 3265
MDL No.: MFCD00000856
Formel:
C₁₃H₁₀Cl₂ Kogepunkt: 159…160 °C (2 mmHg) Massefylde: 1,239 Flammepunkt: 194 °C (381 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000856 CAS-nummer: 134-83-8 EINECS: 205-158-6 UN nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...