Bromcresol lilla, pure indicator grade
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
151330050 151330250 151331400 151330010
ACRO151330050EA 352 DKK
ACRO151330050 ACRO151330250 ACRO151331400 ACRO151330010
Bromcresol lilla, pure indicator grade
Bromcresol lilla
Formel: C₂₁H₁₆Br₂O₅S
Molekylvægt: 540.23 g/mol
Smeltepunkt: 245 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011681
CAS-nummer: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371
Bestil nu
Specifikationer Testresultater
Appearance Light pink to pink-orange Powder
Infrared spectrum Conforms
Solubility (0.1 % in 50 % ethanol) Clear
UV (20 mg/l in methanol) Complies
Visual transition interval from pH 5.2 (yellow) to pH 6.8 (purple)