1-Propyl-4-ethynylbenzen ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne

B25065.06 B25065.14
B25065.06EA 1070 DKK
B25065.06 B25065.14
1-Propyl-4-ethynylbenzen ≥97%
1-Propyl-4-ethynylbenzen

Purity: 97% CAS No.: 62452-73-7 Molecular Formula: C11H12 Formula Weight: 144.22 Flash Point: 67°(153°F) Refractive Index: 1.5340 Boiling Point: 205° MDL No.: MFCD00173886

Formel: C₁₁H₁₂
Kogepunkt: 205 °C
Massefylde: 0,91
Flammepunkt: 67 °C (153 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00173886
CAS-nummer: 62452-73-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR