Cell Locker™ system for Heracell VIOS 160i ™
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50148173 50154739 50153148 50154739X6 50151650X6 50151650
HERA50148173EA 3970 DKK
HERA50148173 390-1047 390-1046 390-1048 390-1045 390-1044
Cell Locker™ system for Heracell VIOS 160i ™
Inkubatorer CO2-inkubatorer
Compatible models of the Heracell VIOS 160i, with modified chambers, can be customised with cell lockers. These autoclavable, polycarbonate cell lockers chambers have been designed for improved culturing efficiency and security for sensitive cultures, such as stem and primary cells, in frequently opened or shared use incubators.
  • Minimise the effect of routine door openings and reduce variability between cell lockers
  • Organise cultures by separating multiple users, cell types, samples or projects within one shared incubator space
  • Prevent cross contamination