2-Methylhydroquinon 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
150830010
150832500
150830050
ACRO150830010EA
1620
DKK
ACRO150830010
ACRO150832500
ACRO150830050
2-Methylhydroquinon 99%
2-Methylhydroquinon
Formel:
C₇H₈O₂ Kogepunkt: 285 °C (1013 hPa) Smeltepunkt: 126 °C Flammepunkt: 172 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002345 CAS-nummer: 95-71-6 EINECS: 202-443-7 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to light beige to light grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | ≥126 °C |
GC | ≥98.5 % |
Ash | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...