2-Methyl-1-phenyl-2-propanol ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98+%
CAS No.: 100-86-7
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 82°(179°F)
Refractive Index: 1.5150
Melting Point: 23-25°
Boiling Point: 215°
Beilstein Reference: 1855608
MDL No.: MFCD00004465
Formel:
C₆H₅CH₂C(CH₃)(OH)CH₃ Kogepunkt: 215 °C Smeltepunkt: 23…25 °C Massefylde: 0,974 Flammepunkt: 82 °C (179 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004465 CAS-nummer: 100-86-7 EINECS: 202-896-0 |