3,4-Dimethoxybenzylalkohol ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol
A13396.14
A13396.22
A13396.36
A13396.14EA
483
DKK
A13396.14
A13396.22
A13396.36
3,4-Dimethoxybenzylalkohol ≥97%
3,4-Dimethoxybenzylalkohol
Purity: 97%
CAS No.: 93-03-8
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point:
Formel:
C₉H₁₂O₃ Kogepunkt: 297…298 °C Smeltepunkt: ∼ 22 °C Massefylde: 1,176 Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004638 CAS-nummer: 93-03-8 EINECS: 202-212-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...