VWR®, Petri Dishes, Glass
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
391-0576EA 629 DKK
391-0576 391-0577 391-0578 391-0579 391-0580 391-0581
VWR®, Petri Dishes, Glass
Dyrkningsplader Petriskåle
Made from borosilicate 3.3 glass that can, therefore, be sterilised and/or used for applications that require heat. These reusable glass Petri dishes are a green alternative to Petri dishes made from plastic, ideal for smaller laboratories.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable and sterilisable