6-Fluor-m-cresol ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Fluoro-5-methylphenol, 1-Fluoro-2-hydroxy-4-methylbenzene
B20275.03
B20275.06
B20275.03EA
423
DKK
B20275.03
B20275.06
6-Fluor-m-cresol ≥97%
6-Fluor-m-cresol
Purity: 97%
CAS No.: 63762-79-8
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 67°(152°F)
Refractive Index: 1.5105
Boiling Point: 172-174°
Beilstein Reference: 2433017
UN No.: 3265
MDL No.: MFCD00190101
Formel:
C₇H₇FO Kogepunkt: 172…174 °C Massefylde: 1,155 Flammepunkt: 67 °C (152 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00190101 CAS-nummer: 63762-79-8 UN nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...