5-Fluor-o-toluidin 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
119460250
119461000
ACRO119460250EA
964
DKK
ACRO119460250
ACRO119461000
5-Fluor-o-toluidin 99%
5-Fluor-o-toluidin
Formel:
C₇H₈FN Molekylvægt: 125.15 g/mol Kogepunkt: 98…100 °C (15 mmHg) Smeltepunkt: 38…40 °C Massefylde: 1.13 Flammepunkt: 90 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007764 CAS-nummer: 367-29-3 EINECS: 206-689-6 UN nummer: 2811 ADR: 6.1 II |
Specifikationer Testresultater
Appearance | Brown to purple Crystalline solid or liquid (May darken on storage) |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...