4-(Trifluormethyl)benzoesyrechlorid ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-(Trifluoromethyl)benzoyl chloride, 4-(Trifluoromethyl)benzoylchloride, 4-(Trifluoromethyl)benzene-1-carbonyl chloride

A14176.06 A14176.14
A14176.06EA 793 DKK
A14176.06 A14176.14
4-(Trifluormethyl)benzoesyrechlorid ≥97%
4-(Trifluormethyl)benzoesyrechlorid

Purity: 98% CAS No.: 329-15-7 Molecular Formula: C8H4ClF3O Formula Weight: 208.57 Flash Point: 78°(172°F) Refractive Index: 1.4770 Odour: Pungent Melting Point: -3° Boiling Point: 188-190° Beilstein Reference: 391282 UN No.: 3265 MDL No.: MFCD00000694

Formel: C₈H₄ClF₃O
Molekylvægt: 208.57 g/mol
Kogepunkt: 188…190 °C
Smeltepunkt: –3 °C
Massefylde: 1.404
Flammepunkt: 78 °C (172 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000694
CAS-nummer: 329-15-7
EINECS: 206-342-9
UN nummer: 3265
ADR: 8 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR