4'-Aminoacetophenon 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
103090100
103090250
103091000
103095000
ACRO103090100EA
177
DKK
ACRO103090100
ACRO103090250
ACRO103091000
ACRO103095000
4'-Aminoacetophenon 99%
4'-Aminoacetophenon
Formel:
C₈H₉NO Kogepunkt: 293 °C (760 mmHg) Smeltepunkt: 96…102 °C Massefylde: 1,13 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007896 CAS-nummer: 99-92-3 EINECS: 202-801-2 Merck Index: 13,00412 |
Specifikationer Testresultater
Appearance | Light yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 104 to 107 °C |
GC | ≥98.5 % |
Absorption Ratio P-15/P+15 | 1.16 to 1.35 |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...