Benzoylisothiocyanat 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
314630050
314630500
ACRO314630050EA
398
DKK
ACRO314630050
ACRO314630500
Benzoylisothiocyanat 98%
Benzoylisothiocyanat
Formel:
C₈H₅NOS Kogepunkt: 128…131 °C (15 mmHg) Smeltepunkt: –1 °C Massefylde: 1,21 g/cm³ Flammepunkt: >110 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00004815 CAS-nummer: 532-55-8 EINECS: 208-540-0 UN nummer: 2810 ADR: 6.1,III Merck Index: 13,01115 |
Specifikationer Testresultater
Appearance | Clear yellow to orange-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6350 to 1.6385 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...