2-Acetylbenzoesyre ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A10216.09
A10216.18
A10216.30
A10216.09EA
539
DKK
A10216.09
A10216.18
A10216.30
2-Acetylbenzoesyre ≥98%
2-Acetylbenzoesyre
Purity: 98+%
CAS No.: 577-56-0
Molecular Formula: C9H8O3
Formula Weight: 164.16
Odour: Odourless
Melting Point: 115-118°
Beilstein Reference: 1210557
MDL No.: MFCD00002475
Formel:
C₉H₈O₃ Kogepunkt: 110…112 °C (2 mmHg) Smeltepunkt: 115…118 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002475 CAS-nummer: 577-56-0 EINECS: 209-413-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...