Safety shoes, lace-up, suXXeed® Offroad S3, Heckel
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6273342 6273343 6273336 6273337 6273346 6273347 6273348 6273339 6273344 6273345 6273338 6273340 6273341
UVEX6273342EA 0 DKK
UVEX6273342 UVEX6273343 UVEX6273336 UVEX6273337 UVEX6273346 UVEX6273347 UVEX6273348 UVEX6273339 UVEX6273344 UVEX6273345 UVEX6273338 UVEX6273340 UVEX6273341
Safety shoes, lace-up, suXXeed® Offroad S3, Heckel
Sko og støvler
Metal-free, flexible and low weight safety ankle boot made from water-resistant oiled grain leather, microfiber inserts, water-resistant tongue and collar, reflective elements, non-metallic eyelets and hooks and a double density PU2D sole resistant to chemicals and hydrocarbons.
  • Quick drying, highly breathable 3D mesh lining
  • Anatomically shaped hygienic insole
  • EVA shock absorber heel
  • Excellent grip and selfcleaning profile
  • Injection moulded PU scuffcap
  • Metal-free protective toecap (200 J)
  • Metal-free puncture resistant midsole
Suitable for building and construction, general industry, energy, logistic, maintenance, utilities, damp working conditions.
Colour: Brown
Certificeringer: EN ISO 20345: 2011; S3, CI, SRC.