3'-Brompropiophenon ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
m-Bromopropiophenone, 1-(3-Bromophenyl)propan-1-one
A16757.06
A16757.14
A16757.06EA
452
DKK
A16757.06
A16757.14
3'-Brompropiophenon ≥97%
3'-Brompropiophenon
Purity: 97%
CAS No.: 19829-31-3
Molecular Formula: C9H9BrO
Formula Weight: 213.08
Flash Point:
Formel:
C₉H₉BrO Kogepunkt: 144…145 °C (20 mmHg) Smeltepunkt: 38…41 °C Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000084 CAS-nummer: 19829-31-3 EINECS: 243-353-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...